missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Anhydrous, Pellets, FCC, 93-100.5%, Spectrum™ Chemical

Catalog No. 18601254
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601254 500 g Poly Bottle
18601255 2.5 kg Poly Pail
18601256 12 kg Poly Pail
18601257 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601254 Supplier Spectrum Chemical Mfg Cor Supplier No. C1977500GM
Only null left

C1977, 10043-52-4, CaCl2

Spectrum™ Chemical Calcium Chloride, Anhydrous, Pellets, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 93 to 100.5%
Grade FCC
Packaging Poly Bottle