missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Anhydrous, granular, Solstice

Catalog No. 6003023
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6003023 100 g
6003025 500 g
6003024 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6003023 Supplier Solstice Supplier No. C1016100G
Only null left

Anhydrous, granular, ≤7.0 mm,≥93.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium;dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Quantity 100 g
Linear Formula CaCl2
UN Number NONH for all modes of transport
Formula Weight 110.98g/mol
Percent Purity ≥93%
Vapor Pressure 0.01 mmHg (20°C)
Chemical Name or Material Calcium chloride
Hazard Statement P305 + P351 + P338
RTECSNumber EV9800000