missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Anhydrous, Granular, FCC, 93-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Calcium Chloride, Anhydrous, Granular, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| pH | 7 |
| CAS Max % | 100% |
| Quantity | 500 g |
| Percent Purity | 93 to 100.5% |
| Grade | FCC |
| Packaging | Poly Bottle |