missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Anhydrous, Desiccant, 4 Mesh, Approx. 95%, Spectrum™ Chemical

Catalog No. 18601240
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601240 2.5 kg Poly Pail
18601239 6 x 500 g Amber Glass Bottle
18601241 4 x 2.5 kg Poly Pail
18601242 12 kg Poly Pail
18601243 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601240 Supplier Spectrum Chemical Mfg Cor Supplier No. C10752.5KG

C1075, 10043-52-4, CaCl2

Spectrum™ Chemical Calcium Chloride, Anhydrous, Desiccant, 4 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 100%
Quantity 2.5 kg
Packaging Poly Pail
Percent Purity ∼95%
Grade Reagent
Chemical Name or Material Calcium Chloride, Anhydrous, Desiccant