missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, ∽97%, MP Biomedicals
Specifications
Specifications
| Quantity | 1 kg |
| Assay Percent Range | ∽97% |
| Formula Weight | 111 |
| Chemical Name or Material | Calcium Chloride, Anhydrous |
Safety and Handling
| Recommended Storage | Room Temperature |