missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, ∽97%, MP Biomedicals

Catalog No. ICN15350290
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15350290 500 g
ICN15350291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15350290 Supplier MP Biomedicals Inc Supplier No. 0215350290
Only null left

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Quantity 500 g
Assay Percent Range ∽97%
Formula Weight 111
Chemical Name or Material Calcium Chloride, Anhydrous
Recommended Storage Room Temperature