missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 96%, for analysis, granules

Catalog No. AC219171000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219171000 100 g Plastic bottle
AC219175000 500 g Plastic bottle
AC219170025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC219171000 Supplier Thermo Scientific Chemicals Supplier No. 219171000

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point >1600.0°C
Melting Point 782.0°C
Physical Form Granules
Quantity 100 g
Assay Percent Range 95% min. (Complexometry)
Linear Formula CaCl2
Fieser 01,104; 04,368
Merck Index 15, 1661
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
Formula Weight 110.99
Percent Purity 96%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium chloride, Granules
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-140-8

RUO – Research Use Only