missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, ≥96%, (ACS Reagent Grade), MP Biomedicals
Desiccant
Supplier: MP Biomedicals Inc 0219381980
Specifications
| Calcium Chloride | |
| 100 g | |
| CaCl2 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| calcium dichloride | |
| 5284359 | |
| 111 |
| 10043-52-4 | |
| ≥96% | |
| calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
| [Cl-].[Cl-].[Ca++] | |
| 110.98 | |
| CHEBI:3312 | |
| ACS Reagent |
Chemical Identifiers
| 10043-52-4 | |
| 110.98 | |
| calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
| CHEBI:3312 | |
| [Cl-].[Cl-].[Ca++] |
| CaCl2 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| 5284359 | |
| calcium dichloride |
Safety and Handling
Recommended Storage : Room Temperature