missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, ≥96%, (ACS Reagent Grade), MP Biomedicals
Specifications
Specifications
| Quantity | 1 kg |
| Assay Percent Range | ≥96% |
| Formula Weight | 111 |
| Grade | ACS Reagent |
| Chemical Name or Material | Calcium Chloride |
Safety and Handling
| Recommended Storage | Room Temperature |