missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, ≥96%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19381950
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19381950 50 g
ICN19381980 100 g
ICN19381990 500 g
ICN19381991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19381950 Supplier MP Biomedicals Inc Supplier No. 0219381950

Desiccant

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Quantity 50 g
Assay Percent Range ≥96%
Formula Weight 111
Grade ACS Reagent
Chemical Name or Material Calcium Chloride
Recommended Storage Room Temperature