missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, 2.75% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 172032
Specifications
| Calcium Chloride | |
| 10035-04-8,7732-18-5 | |
| 98.0,3.59 | |
| Laboratory | |
| Aqueous Solution | |
| 1 L | |
| 2.75% (w/v) |
| Colorless | |
| 95.0,3.45 | |
| 2.75% | |
| Natural Poly Bottle | |
| 8 to 9 | |
| Liquid |
Chemical Identifiers
| 10035-04-8 |