missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, Low in Alkalis, Powder, ACS, 99%, Spectrum™ Chemical
$372.50 - $10742.33
Chemical Identifiers
| CAS | 471-34-1 |
|---|---|
| Molecular Formula | CCaO3 |
| Molecular Weight (g/mol) | 100.09 |
| MDL Number | MFCD00010906 |
| InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
| IUPAC Name | calcium carbonate |
| SMILES | [Ca++].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18609440
|
Spectrum Chemical Mfg Cor
C1071125GM |
125 g | Amber Glass Bottle |
Each for $372.50
|
|
||||
|
18609441
|
Spectrum Chemical Mfg Cor
C1071500GM |
500 g | Amber Glass Bottle |
Each for $995.90
|
|
||||
|
18609442
|
Spectrum Chemical Mfg Cor
C107125KG |
2.5 kg | Poly Pail |
Each for $2,157.50
|
|
||||
|
18609443
|
Spectrum Chemical Mfg Cor
C107145KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.