missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, Low in Alkalis, Powder, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C1071500GM

 View more versions of this product

Catalog No. 18609441


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

471-34-1
500 g
MFCD00010906
[Ca++].[O-]C([O-])=O
100.09
ACS
1
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
99%
Amber Glass Bottle

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
calcium carbonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.