missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, Light Powder, USP, 98-100.5%, Spectrum™ Chemical
CA130, 471-34-1, CaCO3
$547.79 - $4710.04
Chemical Identifiers
| CAS | 471-34-1 |
|---|---|
| Molecular Formula | CCaO3 |
| Molecular Weight (g/mol) | 100.09 |
| MDL Number | MFCD00010906 |
| InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
| IUPAC Name | calcium carbonate |
| SMILES | [Ca++].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601227
|
Spectrum Chemical Mfg Cor
CA130500GM |
500 g | Poly Bottle |
Each for $547.79
|
|
||||
|
18601228
|
Spectrum Chemical Mfg Cor
CA13025KG |
2.5 kg | Poly Pail |
Each for $1,595.50
|
|
||||
|
18601229
|
Spectrum Chemical Mfg Cor
CA13012KG |
12 kg | Fiber Drum |
Each for $2,421.39
|
|
||||
|
18601231
|
Spectrum Chemical Mfg Cor
CA13025KGBL |
25 kg | Fiber Drum |
Each for $4,551.82
|
|
||||
|
18601230
|
Spectrum Chemical Mfg Cor
CA13045KGBL |
45 kg | Fiber Drum |
Each for $4,710.04
|
|
||||