missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, Light Powder, USP, 98-100.5%, Spectrum™ Chemical

Catalog No. 18601227
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601227 500 g Poly Bottle
18601228 2.5 kg Poly Pail
18601229 12 kg Fiber Drum
18601231 25 kg Fiber Drum
18601230 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601227 Supplier Spectrum Chemical Mfg Cor Supplier No. CA130500GM

CA130, 471-34-1, CaCO3

Spectrum™ Chemical Calcium Carbonate, Light Powder, USP is used as a calcium supplement and as an antacid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 2%
Packaging Poly Bottle