missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium bromide hydrate, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 730.0°C |
| Quantity | 250 g |
| Packaging | Plastic bottle |
| Linear Formula | CaBr2·xH2O |
| Solubility Information | (10% in water) Clear colorless to slightly hazy |
| Formula Weight | 199.9 |
| Grade | Extra Pure |
| Chemical Name or Material | Calcium bromide hydrate |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
RUO – Research Use Only