accessibility menu, dialog, popup

UserName

CAS RN 71626-99-8

IUPAC Name: calcium dibromide
Synonyms: calcium bromide|CaBr2|calcium dibromide|kalziumbromid
Molecular Weight (g/mol): 199.89
Molecular Formula: Br2Ca
InChi Key: WGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES: [Ca++].[Br-].[Br-]
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)
Grade 
  • (3)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)