missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, for analysis

Catalog No. p-46008
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300845000 500 g Plastic bottle
AC300840025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC300845000 Supplier Thermo Scientific Chemicals Supplier No. 300845000
Only null left

CAS: 71626-99-8 | Br2Ca | 199.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Melting Point 730.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 500 g
Linear Formula CaBr2·xH2O
Solubility Information Solubility in water: 1420g/L (20°C). Other solubilities: soluble in methanol, ethanol
Formula Weight 199.9
Percent Purity ≥83.0% (Complexometry) (CaBr2)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium bromide hydrate, For analysis
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only