missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, 95%

Catalog No. p-4407310
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1882006 5 g
AA1882022 100 g
AA1882036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1882006 Supplier Thermo Scientific Chemicals Supplier No. 1882006
Only null left

CAS: 71626-99-8 | Br2Ca | 199.89 g/mol

Calcium bromide hydrate is used as freezing mixtures, food preservatives and photography. Its aqueous solution is used in drilling fluids and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 95.0%
Solubility Information Soluble in water.
Formula Weight 199.90 (Anhydrous)
Odor Odorless
Appearance White powder
Chemical Name or Material Calcium bromide hydrate
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 232-164-6
RTECSNumber EV9328000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only