missing translation for 'onlineSavingsMsg'
Learn More

Calcium benzoate hydrate, 98%, water <8%

Catalog No. aaa1235836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1235836 500 g
AAA1235822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1235836 Supplier Thermo Scientific Chemicals Supplier No. A1235836
Only null left

CAS: 2090-05-3 | C19H20O4 | 312.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium benzoate hydrate is used in used in the food industry as a preservative.

Solubility
Soluble in water (2.6 g/100ml at 0°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2090-05-3

Specifications

Molecular Formula C14H10CaO4
MDL Number MFCD09953675
Quantity 500 g
Synonym calcium benzoate, calcium dibenzoate, unii-3qde968mkd, 3qde968mkd, benzoic acid, calcium salt, benzoic acid calcium, acmc-20alj1, calcium 2+ dibenzoate, calcium benzoate, reagent, e213
benzoic acid
Solubility Information Soluble in water (2.6g/100ml at 0°C).
InChI Key NSQPPSOSXWOZNH-UHFFFAOYSA-L
SMILES [Ca++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1
IUPAC Name calcium;dibenzoate
Molecular Weight (g/mol) 282.31
PubChem CID 62425
Formula Weight 282.32 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Calcium benzoate hydrate, Water <8%
Show More Show Less
EINECSNumber 218-235-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only