missing translation for 'onlineSavingsMsg'
Learn More

Calcium Alginate, TCI America™

Catalog No. p-7117036
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A073825G 25 g
A0738500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A073825G Supplier TCI America Supplier No. A073825G
Only null left

Chemical Identifiers

CAS 9005-35-0
Molecular Formula C18H24CaO19
Molecular Weight (g/mol) 584.449
MDL Number MFCD00143567
InChI Key OKHHGHGGPDJQHR-YMOPUZKJSA-L
Synonym Alginic Acid Calcium Salt
PubChem CID 44630049
IUPAC Name calcium;(2S,3S,4S,5S,6R)-6-[(2R,3S,4R,5S,6R)-2-carboxy-6-[(2R,3S,4R,5S,6R)-2-carboxylato-4,5,6-trihydroxyoxan-3-yl]oxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylate
SMILES C1(C(C(OC(C1O)OC2C(C(C(OC2C(=O)O)OC3C(C(C(OC3C(=O)[O-])O)O)O)O)O)C(=O)[O-])O)O.[Ca+2]

Specifications

Physical Form Crystalline Powder
Quantity 25 g
Chemical Name or Material Calcium Alginate
EINECSNumber (8)-0237
RTECSNumber AZ5810000
TSCA Yes