accessibility menu, dialog, popup

UserName

OOHOOOOHOOOOOHOHOHHOHOOOHOOHCa

CAS RN 9005-35-0

IUPAC Name: calcium 6-({2-carboxy-6-[(2-carboxylato-4,5,6-trihydroxyoxan-3-yl)oxy]-4,5-dihydroxyoxan-3-yl}oxy)-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms: calcium 6-({2-carboxy-6-[(2-carboxylato-4,5,6-trihydroxyoxan-3-yl)oxy]-4,5-dihydroxyoxan-3-yl}oxy)-3,4,5-trihydroxyoxane-2-carboxylate calcium alginate
Molecular Weight (g/mol): 584.45
Molecular Formula: C18H24CaO19
InChi Key: OKHHGHGGPDJQHR-UHFFFAOYNA-L
SMILES: [Ca++].OC1OC(C(OC2OC(C(OC3OC(C(O)C(O)C3O)C([O-])=O)C(O)C2O)C(O)=O)C(O)C1O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results