missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014931
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014931 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014931 Supplier Merck Emd Millipore Supplier No. PHR13625G
Only null left
Calcium Acetate belongs to the group of calcium salts, widely used as phosphorus binders in patients with chronic renal failure.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 62-54-4
Physical Form Neat
Linear Formula (CH3 CO2)2 Ca
MDL Number MFCD00012448
Synonym Calcium acetate
Type Certified Reference Material
Quantity 5 g
Formula Weight 158.17
Grade Certified Reference Material