missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate, Monohydrate, Powder, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C106525KG

 View more versions of this product

Catalog No. 18609434


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5743-26-0
2.5 kg
XQKKWWCELHKGKB-UHFFFAOYSA-L
calcium diacetate hydrate
99%
Poly Pail
1
C4H8CaO5
O.[Ca++].CC([O-])=O.CC([O-])=O
176.18
ACS

Chemical Identifiers

5743-26-0
176.18
calcium diacetate hydrate
C4H8CaO5
XQKKWWCELHKGKB-UHFFFAOYSA-L
O.[Ca++].CC([O-])=O.CC([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.