missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate, Monohydrate, Powder, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165274
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609432 125 g Amber Glass Bottle
18609433 500 g Amber Glass Bottle
18609434 2.5 kg Poly Pail
18609435 12 kg Fiber Drum
18609436 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609432 Supplier Spectrum Chemical Mfg Cor Supplier No. C1065125GM
Only null left

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.18
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
IUPAC Name calcium diacetate hydrate
SMILES O.[Ca++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.