missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis)

Catalog No. p-7049125
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1297406 5 g
AA1297414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1297406 Supplier Thermo Scientific Chemicals Supplier No. 01297406
Only null left

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

Calcium acetate has been utilized in the crystallization of proteins, such as human annexin II and the bacterial heme oxygenase (Hmu O), and of a complex of human CDK6 with a virus-encoded cyclin. It has been used in the manufactureof metallic soaps and synthetic resins, in lubricants as a corrosion inhibitor, and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 5 g
Beilstein 3692527
Merck Index 14,1646
Solubility Information Soluble in water. Slightly soluble in methanol,hydrazine insoluble in acetone,ethanol and benzene.
Formula Weight 158.17 (Anhydrous)
Percent Purity 99.9965%
Grade Puratronic
Sensitivity Hygroscopic
Density 1.5 g/mL
Assay (metals basis)
Chemical Name or Material Calcium acetate hydrate
Show More Show Less
EINECSNumber 200-540-9
RTECSNumber AF7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only