missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, 99%

Catalog No. AAA1494736
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1494736 500 g
AAA1494722 100 g
AAA149470E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1494736 Supplier Thermo Scientific Chemicals Supplier No. A1494736

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Density 1.5
Linear Formula Ca(OOCCH3)2·xH2O
Quantity 500 g
Beilstein 3692527
Merck Index 14,1646
Solubility Information Soluble in water.
Formula Weight 158.17 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Calcium acetate hydrate
EINECSNumber 200-540-9
RTECSNumber AF7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only