missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, 99%, extra pure

Catalog No. p-4509929
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211052500 250 g Plastic bottle
AC211050010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211052500 Supplier Thermo Scientific Chemicals Supplier No. 211052500
Only null left

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
Synonym Acetic acid, calcium salt hydrate
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Color White
pH 7.0 to 9.0 (10% soln., 20°C)
Physical Form Powder and/or Agglomerates
Quantity 250 g
Assay Percent Range 98.5 to 101.5% (on anhydrous substance) (Complexometry)
Linear Formula (CH3CO2)2Ca
Beilstein 02,IV,113
Fieser 13,60; 15,46
Merck Index 14,1646
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 158.17
Percent Purity 99%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Calcium acetate hydrate
Show More Show Less

RUO – Research Use Only