missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, 99%, for analysis

Catalog No. AC447860010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447860010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC447860010 Supplier Thermo Scientific Chemicals Supplier No. 447860010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
Synonym Acetic acid, calcium salt hydrate
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 98.5% min. (on dried substance) (Complexometry)
Linear Formula (CH3CO2)2Ca
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 158.17
Percent Purity 99%
Grade Analytical
Chemical Name or Material Calcium acetate hydrate

RUO – Research Use Only