missing translation for 'onlineSavingsMsg'
Learn More

Calcium 4-Methyl-2-oxovalerate Hydrate 98.0+%, TCI America™

Catalog No. K00235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00235G 5 g
K002325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00235G Supplier TCI America Supplier No. K00235G
Only null left

Chemical Identifiers

CAS 51828-95-6
Molecular Formula C12H20CaO6
Molecular Weight (g/mol) 300.364
MDL Number MFCD00004163
InChI Key QSTGQEJKDFGVPP-UHFFFAOYSA-N
Synonym 4-Methyl-2-oxovaleric Acid Calcium Salt
PubChem CID 131854617
IUPAC Name calcium;4-methyl-2-oxopentanoic acid
SMILES CC(C)CC(=O)C(=O)O.CC(C)CC(=O)C(=O)O.[Ca]

Specifications

Color White
Quantity 5 g
Formula Weight 298.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium 4-Methyl-2-oxovalerate Hydrate