accessibility menu, dialog, popup

UserName

CAS RN 51828-95-6

CaH3CCH3OOO

CAS RN 51828-95-6

IUPAC Name: calcium 4-methyl-2-oxopentanoate
Synonyms: calcium α-ketoisocaproate
Molecular Weight (g/mol): 169.21
Molecular Formula: C6H9CaO3
InChi Key: IHROUNIARQTAPN-UHFFFAOYSA-M
SMILES: [Ca++].CC(C)CC(=O)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results