missing translation for 'onlineSavingsMsg'
Learn More

Calcium 3-Methyl-2-oxovalerate Hydrate 98.0+%, TCI America™

Supplier:  TCI America K002225G

 View more versions of this product

Catalog No. K002225G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium 3-Methyl-2-oxovalerate Hydrate
White
MFCD00002583
3-Methyl-2-oxovaleric Acid Calcium Salt
CCC(C)C(=O)C(=O)[O-].CCC(C)C(=O)C(=O)[O-].[Ca+2]
298.348
298.35
Crystalline Powder
66872-75-1
C12H18CaO6
25 g
PTFSVYLXDCGPFY-UHFFFAOYSA-L
calcium;3-methyl-2-oxopentanoate
6099313
≥98.0% (T)

Chemical Identifiers

66872-75-1
298.348
PTFSVYLXDCGPFY-UHFFFAOYSA-L
6099313
CCC(C)C(=O)C(=O)[O-].CCC(C)C(=O)C(=O)[O-].[Ca+2]
C12H18CaO6
MFCD00002583
3-Methyl-2-oxovaleric Acid Calcium Salt
calcium;3-methyl-2-oxopentanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions