Learn More
Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)
CAS: 136-51-6 | C16H30CaO4 | 326.49 g/mol
$82.70 - $235.03
Chemical Identifiers
| CAS | 136-51-6 |
|---|---|
| Molecular Formula | C16H30CaO4 |
| Molecular Weight (g/mol) | 326.49 |
| MDL Number | MFCD00014001 |
| InChI Key | LTPCXXMGKDQPAO-UHFFFAOYSA-L |
| Synonym | calcium 2-ethylhexanoate, calcium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, calcium salt, hexanoic acid, 2-ethyl-, calcium salt 2:1, calcium 2-ethyl-hexanoate, octope ca, calcium hex-cem, nikka octhix calcium, calcium 2-ethylhexoate, dsstox_cid_7063 |
| PubChem CID | 61082 |
| IUPAC Name | calcium;2-ethylhexanoate |
| SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ca+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 136-51-6 | |
| 326.49 | |
| LTPCXXMGKDQPAO-UHFFFAOYSA-L | |
| 61082 | |
| CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ca+2] |
| C16H30CaO4 | |
| MFCD00014001 | |
| calcium 2-ethylhexanoate, calcium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, calcium salt, hexanoic acid, 2-ethyl-, calcium salt 2:1, calcium 2-ethyl-hexanoate, octope ca, calcium hex-cem, nikka octhix calcium, calcium 2-ethylhexoate, dsstox_cid_7063 | |
| calcium;2-ethylhexanoate |
Specifications
| 136-51-6 | |
| C16H30CaO4 | |
| 5 g | |
| LTPCXXMGKDQPAO-UHFFFAOYSA-L | |
| calcium;2-ethylhexanoate | |
| 61082 | |
| Superconductor | |
| Liquid |
| Weak, characteristic | |
| MFCD00014001 | |
| calcium 2-ethylhexanoate, calcium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, calcium salt, hexanoic acid, 2-ethyl-, calcium salt 2:1, calcium 2-ethyl-hexanoate, octope ca, calcium hex-cem, nikka octhix calcium, calcium 2-ethylhexoate, dsstox_cid_7063 | |
| CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ca+2] | |
| 326.49 | |
| 326.5 | |
| 3 to 8% Ca | |
| Calcium 2-ethylhexanoate, in 2-ethylhexanoic acid |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H361d
EINECSNumber : 205-249-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only