missing translation for 'onlineSavingsMsg'
Learn More

Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)

Catalog No. AA3665706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3665706 5 g
AA3665714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3665706 Supplier Thermo Scientific Chemicals Supplier No. 03665706

CAS: 136-51-6 | C16H30CaO4 | 326.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136-51-6
Molecular Formula C16H30CaO4
Molecular Weight (g/mol) 326.49
MDL Number MFCD00014001
InChI Key LTPCXXMGKDQPAO-UHFFFAOYSA-L
Synonym calcium 2-ethylhexanoate, calcium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, calcium salt, hexanoic acid, 2-ethyl-, calcium salt 2:1, calcium 2-ethyl-hexanoate, octope ca, calcium hex-cem, nikka octhix calcium, calcium 2-ethylhexoate, dsstox_cid_7063
PubChem CID 61082
IUPAC Name calcium;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ca+2]

Specifications

Odor Weak, characteristic
Quantity 5 g
Formula Weight 326.5
Grade Superconductor
Concentration or Composition (by Analyte or Components) 3 to 8% Ca
Physical Form Liquid
Chemical Name or Material Calcium 2-ethylhexanoate, in 2-ethylhexanoic acid
Hazard Category H315-H319-H335-H361d
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 205-249-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only