missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Calcein Violet 450 AM Viability Dye
Description
- Molecular weight: 600g/mol
- Peak excitation: 408nm
- Peak emission: 450nm
- Reconstitute in high-quality, freshly opened DMSO
- Once reconstituted, store at −20°C with dessicant and use within a short period of time. Avoid freeze-thawing
- For fluorescent microscopy, detect using the appropriate filter sets
- For flow cytometric analysis, excite with the violet laser line (405nm) and detect using filters for Pacific Blue™/eFluor™ 450 (450/50)
- Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
Specifications
Specifications
| Color | Violet |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Product Line | eBioscience |
| Quantity | 1 mg |
| Emission Wavelength | 408/450 |
For Research Use Only. Not for use in diagnostic procedures.