missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcein (mixture of isomers) [for Fluorometric Determination of Ca], TCI America™
$192.00 - $628.19
Chemical Identifiers
| CAS | 154071-48-4 |
|---|---|
| Molecular Formula | C30H26N2O13 |
| Molecular Weight (g/mol) | 622.54 |
| MDL Number | MFCD00005049,MFCD09750388 |
| InChI Key | DEGAKNSWVGKMLS-UHFFFAOYSA-N |
| Synonym | Fluorexon, Fluorescein Bis(methyliminodiacetic Acid), Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein |
| PubChem CID | 65079 |
| ChEBI | CHEBI:51903 |
| IUPAC Name | 2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid |
| SMILES | OC(=O)CN(CC(O)=O)CC1=C(O)C=C2OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C3(OC(=O)C4=CC=CC=C34)C2=C1 |
Chemical Identifiers
| 154071-48-4 | |
| 622.54 | |
| DEGAKNSWVGKMLS-UHFFFAOYSA-N | |
| 65079 | |
| 2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid |
| C30H26N2O13 | |
| MFCD00005049,MFCD09750388 | |
| Fluorexon, Fluorescein Bis(methyliminodiacetic Acid), Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein | |
| CHEBI:51903 | |
| OC(=O)CN(CC(O)=O)CC1=C(O)C=C2OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C3(OC(=O)C4=CC=CC=C34)C2=C1 |
Specifications
| 154071-48-4 | |
| MFCD00005049,MFCD09750388 | |
| DEGAKNSWVGKMLS-UHFFFAOYSA-N | |
| 2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid | |
| 65079 | |
| 622.54 | |
| Crystal/Powder |
| C30H26N2O13 | |
| Fluorexon, Fluorescein Bis(methyliminodiacetic Acid), Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein | |
| OC(=O)CN(CC(O)=O)CC1=C(O)C=C2OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C3(OC(=O)C4=CC=CC=C34)C2=C1 | |
| 622.54 | |
| CHEBI:51903 | |
| 1 g | |
| Calcein (mixture of isomers) [for Fluorometric Determination of Ca] |
Safety and Handling
TSCA : Yes