missing translation for 'onlineSavingsMsg'
Learn More

Calcein Disodium Salt, Solstice

Catalog No. 60045203
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045203 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60045203 Supplier Solstice Supplier No. 210305G

for complexometry

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 108750-13-6
Molecular Formula C30H25N2NaO13
Molecular Weight (g/mol) 644.521
MDL Number MFCD00082336
InChI Key WFTKXUHBPZFOMD-UHFFFAOYSA-M
PubChem CID 131674686
IUPAC Name sodium;2',7'-bis[[bis(carboxymethyl)amino]methyl]-6'-hydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-olate
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)[O-])O)CN(CC(=O)O)CC(=O)O.[Na+]

Specifications

UN Number NONH for all modes of transport
Formula Weight 666.5g/mol
Packaging Glass Bottle
Color Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Calcein Disodium Salt, For complexometry