missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALCB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149803
Description
CALCB Polyclonal Antibody for Western Blot, ICC/IF, IHC
CGRP induces vasodilation. It dilates a variety of vessels including the coronary, cerebral and systemic vasculature. Its abundance in the CNS also points toward a neurotransmitter or neuromodulator role.Specifications
| CALCB | |
| Polyclonal | |
| Unconjugated | |
| CALCB | |
| Beta-CGRP; beta-type CGRP; CALC2; Calcb; calcitonin 2; calcitonin gene-related peptide 2; Calcitonin gene-related peptide II; calcitonin related polypeptide beta; calcitonin-related polypeptide beta; calcitonin-related polypeptide, beta; CGRP2; CGRP-II | |
| Rabbit | |
| Protein A | |
| RUO | |
| 797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P10092 | |
| CALCB | |
| Recombinant Calcitonin Gene Related Peptide (CGRP) (Met1-Ala127). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |