missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Vitamin B12

Catalog No. 67915GM
Change view
Click to view available options
Vitamin B12; Qty: 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
67915GM Vitamin B12; Qty: 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67915GM Supplier MilliporeSigma™ Supplier No. 67915GM

Used as an antipernicious anemia factor

  • Part of coenzyme needed for the synthesis of proteins, and nucleic acids
  • Used for metabolism of carbohydrates
  • Plays role in synthesis of myelin

Specifications

Content And Storage 15°C to 30°C
Additive Vitamin B12
Quantity 5 g