Learn More
MilliporeSigma™ Calbiochem™ trans-Retinoic Acid
Catalog No. 554720500MG
Change view
Click to view available options
trans-Retinoic Acid; Qty: 500mg
Catalog No.
554720500MG trans-Retinoic Acid; Qty: 500mg
1 options
Catalog No. 554720500MG Supplier MilliporeSigma™ Supplier No. 554720500MG
Acts as potent modulator of growth and differentiation
  • Inhibits melanocyte adhesion, motility, and growth
  • Has keratolytic activity
  • Induces differentiation of tumor cells of neural origin
  • Induces apoptosis in acute promyelocytic carcinoma cells
Specifications
Content And Storage 2°C to 8°C
Additive trans-Retinoic Acid
Quantity 500 mg