missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ TPEN
Description
- Auxiliary reagent in certain techniques used for measuring free Ca2+ levels in cells
- Also useful in extracting Aβ amyloid deposits from Alzheimer’s disease brain tissue
- Has extremely high affinity for heavy metals but low affinity for Ca2+ and Mg2+
- Induces DNA fragmentation when chelated with Zn2+ and induces apoptosis in thymocytes
- Strips heavy metals from liposome-trapped QUIN 2
Specifications
Specifications
| CAS | 16858-02-9 |
| Physical Form | Solid |
| Synonym | N,N,Nʹ,Nʹ-tetrakis-(2-Pyridylmethyl)ethylenediamine |
| Solubility | DMSO, EtOH, PBS |
| Type | Calbiochem TPEN |
| Quantity | 100 mg |
| Percent Purity | ≥97% (Titration) |