missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Tetrahydrouridine

Catalog No. 58422210MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58422210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58422210MG Supplier MilliporeSigma™ Supplier No. 58422210MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potent competitive inhibitor of cytidine deaminase

Potent competitive inhibitor of cytidine deaminase

Specifications

CAS 18771-50-1
Physical Form Solid
Molecular Formula C9H16N2O6
Color White to off-white
Quantity 10 mg
Formula Weight 248.2 g/mol
Percent Purity ≥90% (TLC)