missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Spermine, Tetrahydrochloride

Catalog No. 56775GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56775GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56775GM Supplier MilliporeSigma™ Supplier No. 56775GM
Only null left

Plays an important role in the regulation of cellular proliferation and differentiation

  • Has both stimulatory and inhibitory effects on NMDA receptors depending on subunit composition of receptor
  • Acts as inhibitor of phospholipase Cα and an activator of phospholipase Cδ
  • Inhibits DNA fragmentation and morphological apoptosis
  • Inhibits neuronal NOS

Specifications

Content And Storage 15-30°C
Form White solid
Product Type Spermine
Quantity 5 g

For Research Use Only