missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ SeaBlock™ Reagent, Salmon Plasma

Catalog No. 558300500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
558300500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 558300500ML Supplier MilliporeSigma™ Supplier No. 558300500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready to use blocking reagent for western blotting immunoassays.

EMD Millipore Calbiochem™ SeaBlock™ Reagent, Salmon Plasma contains salmon plasma proteins that do not interact with mammalian antibodies.

  • Offers reduced background
  • Packaging: Glass Bottle

Specifications

Description SeaBlock Reagent, Salmon Plasma
Quantity 500 mL
Content And Storage 2°C to 8°C