missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Rat Plasma Fibronectin

Catalog No. 341668500UG
Change view
Click to view available options
Quantity:
500μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
341668500UG 500μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 341668500UG Supplier MilliporeSigma™ Supplier No. 341668500UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purified from pooled rat plasma.

EMD Millipore Calbiochem™ Rat Plasma Fibronectin is an effective agent for promoting attachment of cells.

  • Native fibronectin
  • Promotes attachment of cells to commonly-used culture substrates
  • Contaminants: None
  • Detected by IEP using anti-fibronectin anti-rat serum
  • Single Major B by SDS-PAGE
  • CAS: 86088-83-7

Specifications

Concentration In 100mM NaCl, 20mM sodium phosphate buffer, pH 7.3, preservative and reductant-free
Content And Storage −20°C
Quantity 500μg