missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Normal Goat Serum

Used as negative control in any application. MilliporeSigma™; Calbiochem™ Normal Goat Serum is normal carrier sera, produced from healthy, non-immunized goats.

Supplier:  MilliporeSigma™ NS02L1ML

Catalog No. NS02L1ML

Only null left
Explore available promotions

Description

Description

  • Form: Lyophilized
  • Formulation: Lyophilized serum
  • Preservative: ≤0.1% sodium azide
Specifications

Specifications

Ok to freeze. Reconstitute the lypohilized serum in 1 ml sterile distilled H₂O with occasional gentle mixing for at least 2 h at 4°C. Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) with 0.1% azide for short-term storage. Avoid freeze/thaw cycles of solutions.
Lyophilized
1 mL
Yes
Goat
Goat Serum
ELISA, Flow Cytometry, Immunoblotting (Western Blotting), Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Negative control in any application
≤0.1% sodium azide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions