missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Non-Interfering Protein Assay™ Kit

Catalog No. 4882501KTT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4882501KTT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4882501KTT Supplier MilliporeSigma™ Supplier No. 4882501KTT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Overcomes interference of agents found in protein solutions.

EMD Millipore Calbiochem™ Non-Interfering Protein Assay™ Kit is used to precipitate immobilize protein.

  • Overcomes interference of detergents, chelating agents, reducing agents, amines, sugars, urea, others
  • Protein concentration is based on specific binding of copper to peptide backbone
  • Protein concentration increases, the concentration of unbound copper ions decreases
  • Color density is inversely related to the amount of protein present in solution
  • Packaging: Glass Bottle

Specifications

Product Type Non-Interfering Protein Assay Kit
Sufficient For 500 Tests
Kit Contents BSA standard, Color agents A and B, Copper solution I, UPPA™ Reagents I and II, User protocol
Quantity 1 Kit
Storage Requirements 15°C to 30°C