Learn More
MilliporeSigma™ Calbiochem™ NADPH, Tetrasodium Salt
Catalog No. 48197350MG
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
500 mg
1 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48197350MG 50 mg
48197325MG 25 mg
481973100MG 100 mg
481973500MG 500 mg
4819731GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 48197350MG Supplier MilliporeSigma™ Supplier No. 48197350MG
Ubiquitous coenzyme that acts as an electron donor in many reactions utilizing dehydrogenase and reductase enzymes
Specifications
Content And Storage -20°C
Additive NADPH, Tetrasodium Salt
Quantity 50 mg