Learn More
MilliporeSigma™ Calbiochem™ NADPH, Tetrasodium Salt
Ubiquitous coenzyme that acts as an electron donor in many reactions utilizing dehydrogenase and reductase enzymes
Supplier:  MilliporeSigma™ 4819731GM
 View more versions of this product
Catalog No. 4819731GM
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
-20°C
NADPH, Tetrasodium Salt
1 g
Product Suggestions
Product Suggestions