Learn More
MilliporeSigma™ Calbiochem™ NADP, Monosodium Salt
Participates in oxidation-reduction reactions
Supplier: MilliporeSigma™ 481971500MG
Description
- Coenzyme that is widely distributed in living matter
Specifications
| -20°C | |
| NADP, Monosodium Salt | |
| 500 mg |